EMAIL THIS PAGE TO A FRIEND
Journal of lipid research
Macrophage apoAI protects against dyslipidemia-induced dermatitis and atherosclerosis without affecting HDL.
PMID 25593328
Abstract
Tissue cholesterol accumulation, macrophage infiltration, and inflammation are features of atherosclerosis and some forms of dermatitis. HDL and its main protein, apoAI, are acceptors of excess cholesterol from macrophages; this process inhibits tissue inflammation. Recent epidemiologic and clinical trial evidence questions the role of HDL and its manipulation in cardiovascular disease. We investigated the effect of ectopic macrophage apoAI expression on atherosclerosis and dermatitis induced by the combination of hypercholesterolemia and absence of HDL in mice. Hematopoietic progenitor cells were transduced to express human apoAI and transplanted into lethally irradiated LDL receptor(-/-)/apoAI(-/-) mice, which were then placed on a high-fat diet for 16 weeks. Macrophage apoAI expression reduced aortic CD4(+) T-cell levels (-39.8%), lesion size (-25%), and necrotic core area (-31.6%), without affecting serum HDL or aortic macrophage levels. Macrophage apoAI reduced skin cholesterol by 39.8%, restored skin morphology, and reduced skin CD4(+) T-cell levels. Macrophage apoAI also reduced CD4(+) T-cell levels (-32.9%) in skin-draining lymph nodes but had no effect on other T cells, B cells, dendritic cells, or macrophages compared with control transplanted mice. Thus, macrophage apoAI expression protects against atherosclerosis and dermatitis by reducing cholesterol accumulation and regulating CD4(+) T-cell levels, without affecting serum HDL or tissue macrophage levels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PZ0203
Sitaxentan sodium salt, ≥98% (HPLC)
C18H14ClN2NaO6S2