EMAIL THIS PAGE TO A FRIEND
Endocrinology
Excess iodide induces an acute inhibition of the sodium/iodide symporter in thyroid male rat cells by increasing reactive oxygen species.
PMID 25594695
Abstract
Na+/I- symporter (NIS) mediates iodide (I-) uptake in the thyroid gland, the first and rate-limiting step in the biosynthesis of the thyroid hormones. The expression and function of NIS in thyroid cells is mainly regulated by TSH and by the intracellular concentration of I-. High doses of I- for 1 or 2 days inhibit the synthesis of thyroid hormones, a process known as the Wolff-Chaikoff effect. The cellular mechanisms responsible for this physiological response are mediated in part by the inhibition of I- uptake through a reduction of NIS expression. Here we show that inhibition of I- uptake occurs as early as 2 hours or 5 hours after exposure to excess I- in FRTL-5 cells and the rat thyroid gland, respectively. Inhibition of I- uptake was not due to reduced NIS expression or altered localization in thyroid cells. We observed that incubation of FRTL-5 cells with excess I- for 2 hours increased H2O2 generation. Furthermore, the inhibitory effect of excess I- on NIS-mediated I- transport could be recapitulated by H2O2 and reverted by reactive derived oxygen species scavengers. The data shown here support the notion that excess I- inhibits NIS at the cell surface at early times by means of a posttranslational mechanism that involves reactive derived oxygen species.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M32631
2-Methylbutane, ReagentPlus®, ≥99%
C5H12
277258
2-Methylbutane, anhydrous, ≥99%
C5H12
270342
2-Methylbutane, for HPLC, ≥99.5%
C5H12
320404
2-Methylbutane, ReagentPlus®, ≥99%
C5H12
59070
2-Methylbutane, puriss. p.a., ≥99.5% (GC)
C5H12
59060
2-Methylbutane, analytical standard
C5H12
M70800
3-Methyl-1-phenyl-2-pyrazoline-5-one, 99%
C10H10N2O
172553
4,5-Dihydroxy-1,3-benzenedisulfonic acid disodium salt monohydrate, 97%
C6H4Na2O8S2 · H2O
C9322 Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
G7387
Gly-His-Lys acetate salt, BioXtra, γ-irradiated, suitable for cell culture
C16H28N6O6
PHR1428
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
C8H18N2O4S
54461
HEPES, ≥99.0% (T)
C8H18N2O4S
I6634 Insulin from bovine pancreas, powder, BioReagent, suitable for cell culture
C254H377N65O75S6
220051
N-Iodosuccinimide, 95%
C4H4INO2
Y0001138
Phenazone impurity A, European Pharmacopoeia (EP) Reference Standard
C10H10N2O
30315
Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%
IK
60399
Potassium iodide, BioUltra, ≥99.5% (AT)
IK
746428
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
IK
793582
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
IK
03124
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
IK
204102
Potassium iodide, ≥99.99% trace metals basis
IK
P8166
Potassium iodide, suitable for plant cell culture
IK
P2963
Potassium iodide, BioXtra, ≥99.0%
IK
60422
Potassium iodide, tested according to Ph. Eur.
IK
429422
Potassium iodide, anhydrous, beads, −10 mesh, 99.998% trace metals basis
IK
PHR1360
Potassium Iodide, Pharmaceutical Secondary Standard; Certified Reference Material
IK
45656
Rotenone, PESTANAL®, analytical standard
C23H22O6
229865
Selenium, powder, −100 mesh, 99.99% trace metals basis
Se
209651
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Se
209643
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Se