EMAIL THIS PAGE TO A FRIEND
Applied and Environmental Microbiology
Improved production of propionic acid in Propionibacterium jensenii via combinational overexpression of glycerol dehydrogenase and malate dehydrogenase from Klebsiella pneumoniae.
PMID 25595755
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
94425
Propionic acid, analytical standard
C3H6O2