EMAIL THIS PAGE TO A FRIEND
Vaccine
Protective efficacy and immune responses induced by a DNA vaccine encoding codon-optimized PPA1 against Photobacterium damselae subsp. piscicida in Japanese flounder.
PMID 25596456