EMAIL THIS PAGE TO A FRIEND
Journal of Biomaterials Science. Polymer Edition
Optimum design of amphiphilic polymers bearing hydrophobic groups for both cell surface ligand presentation and intercellular cross-linking.
PMID 25597323