EMAIL THIS PAGE TO A FRIEND
Journal of Dairy Science
Cow-to-cow variation in fibroblast response to a toll-like receptor 2/6 agonist and its relation to mastitis caused by intramammary challenge with Staphylococcus aureus.
PMID 25597966