EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Electrogenic and nonelectrogenic ion fluxes across lipid and mitochondrial membranes mediated by monensin and monensin ethyl ester.
PMID 25600660
Abstract
Monensin is a carrier of cations through lipid membranes capable of exchanging sodium (potassium) cations for protons by an electroneutral mechanism, whereas its ethyl ester derivative ethyl-monensin is supposed to transport sodium (potassium) cations in an electrogenic manner. To elucidate mechanistic details of the ionophoric activity, ion fluxes mediated by monensin and ethyl-monensin were measured on planar bilayer lipid membranes, liposomes, and mitochondria. In particular, generation of membrane potential on liposomes was studied via the measurements of rhodamine 6G uptake by fluorescence correlation spectroscopy. In mitochondria, swelling experiments were expounded by the additional measurements of respiration, membrane potential, and matrix pH. It can be concluded that both monensin and ethyl-monensin can perform nonelectrogenic exchange of potassium (sodium) ions for protons and serve as electrogenic potassium ion carriers similar to valinomycin. The results obtained are in line with the predictions based on the crystal structures of the monensin complexes with sodium ions and protons (Huczyński et al., Biochim. Biophys. Acta, 1818 (2012) pp. 2108-2119). The functional activity observed for artificial membranes and mitochondria can be applied to explain the activity of ionophores in living systems. It can also be important for studying the antitumor activity of monensin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H1529
8-Hydroxypyrene-1,3,6-trisulfonic acid trisodium salt, ≥96%
C16H7Na3O10S3