EMAIL THIS PAGE TO A FRIEND
Biochemical and Biophysical Research Communications
Midazolam inhibits the formation of amyloid fibrils and GM1 ganglioside-rich microdomains in presynaptic membranes through the gamma-aminobutyric acid A receptor.
PMID 25600806