EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
Multiple assembly mechanisms anchor the KMN spindle checkpoint platform at human mitotic kinetochores.
PMID 25601404
Abstract
During mitosis, the spindle checkpoint senses kinetochores not properly attached to spindle microtubules and prevents precocious sister-chromatid separation and aneuploidy. The constitutive centromere-associated network (CCAN) at inner kinetochores anchors the KMN network consisting of Knl1, the Mis12 complex (Mis12C), and the Ndc80 complex (Ndc80C) at outer kinetochores. KMN is a critical kinetochore receptor for both microtubules and checkpoint proteins. Here, we show that nearly complete inactivation of KMN in human cells through multiple strategies produced strong checkpoint defects even when all kinetochores lacked microtubule attachment. These KMN-inactivating strategies reveal multiple KMN assembly mechanisms at human mitotic kinetochores. In one mechanism, the centromeric kinase Aurora B phosphorylates Mis12C and strengthens its binding to the CCAN subunit CENP-C. In another, CENP-T contributes to KMN attachment in a CENP-H-I-K-dependent manner. Our study provides insights into the mechanisms of mitosis-specific assembly of the checkpoint platform KMN at human kinetochores.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
97622
2-Mercaptoethanol, for HPLC derivatization, ≥99.0% (GC)
C2H6OS
32883
4-tert-Octylphenol monoethoxylate solution, 1 μg/mL in acetone, analytical standard
C16H26O2
32882
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
C16H26O2
05-368
Anti-phospho-Ser/Thr-Pro MPM-2 Antibody, Anti-phospho-Ser/Thr-Pro MPM-2 Antibody is a Mouse Monoclonal Antibody for detection of phospho-Ser/Thr-Pro also known as Mitotic protein #2 & has been tested in ELISA, IHC, IP & WB.
1012772
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
C2H6O
PHR1070
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
C2H6O
03777
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, for molecular biology, ≥99.0% (T)
C14H24N2O10
47072
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
FNa
80373
Fluoride, certified anion standard solution, 1000 mg/kg F-, for ion chromatography, certified by BAM, according to ISO 17025
FNa
PHR1359
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
C10H17N3O6S
Y0000517
Glutathione, European Pharmacopoeia (EP) Reference Standard
C10H17N3O6S
1295607
Glycerin, United States Pharmacopeia (USP) Reference Standard
C3H8O3
PHR1020
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
C3H8O3
49779
Glycerol, tested according to Ph.Eur., anhydrous
C3H8O3
77067
Glycerol, analytical standard
C3H8O3
12146
Glycerol 3 M solution, 3 M
C3H8O3 · xH2O
02739
Imidazole, pharmaceutical impurity standard, ≥95.0% (HPLC)
C3H4N2
1336500
Imidazole, United States Pharmacopeia (USP) Reference Standard
C3H4N2
I0086000
Imidazole, European Pharmacopoeia (EP) Reference Standard
C3H4N2
PHR1180
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
C3H4N2
63031
Magnesium ion solution for ISE, 0.01 M Mg, analytical standard (for ion-selective electrodes)
Cl2Mg
63042
Magnesium ion solution for ISE, 0.1 M Mg, analytical standard (for ion-selective electrodes)
Cl2Mg
O4511
Okadaic acid from Prorocentrum concavum, ≥90% (HPLC), translucent film
C44H68O13
O9381
Okadaic acid from Prorocentrum concavum, 92-100% (HPLC)
C44H68O13
Y0001326
Ondansetron impurity E, European Pharmacopoeia (EP) Reference Standard
C3H4N2
81845
Propidium iodide, ≥94% (HPLC)
C27H34I2N4
450022
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
FNa
PHR1408
Sodium Fluoride, certified reference material
FNa
67414
Sodium fluoride 0.5 M solution
FNa
T1895
Thymidine, powder, BioReagent, suitable for cell culture
C10H14N2O5