EMAIL THIS PAGE TO A FRIEND
Journal of Neuroscience Research
Effects of (R)-(-)-5-methyl-1-nicotinoyl-2-pyrazoline on glutamate transporter 1 and cysteine/glutamate exchanger as well as ethanol drinking behavior in male, alcohol-preferring rats.
PMID 25601490