EMAIL THIS PAGE TO A FRIEND
International Immunopharmacology
Androctonus australis hector venom contributes to the interaction between neuropeptides and mast cells in pulmonary hyperresponsiveness.
PMID 25601496