EMAIL THIS PAGE TO A FRIEND
Clinica Chimica Acta; International Journal of Clinical Chemistry
A fast semi-quantitative LC-MS method for measurement of intact apolipoprotein A-I reveals novel proteoforms in serum.
PMID 25603406