EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Expedient synthesis of tetrasubstituted pyrroles via a copper-catalyzed cascade inter-/intramolecular cyclization of 1,3-enynes carry a nitro group with amines.
PMID 25604620
Abstract
Various tetrasubstituted pyrroles/pyrazoles have been prepared from nitro-substituted 1,3-enynes with aromatic amines/hydrazines via a copper-catalyzed cascade aza-Michael addition, cyclization and aromatization at room temperature. This protocol is also effective for the synthesis of tetrasubstituted pyrazoles in high yields.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
412740
Bis(triphenylphosphine)palladium(II) dichloride
C36H30Cl2P2Pd
208671
Bis(triphenylphosphine)palladium(II) dichloride, 98%
C36H30Cl2P2Pd
741248
Bis(triphenylphosphine)palladium(II) dichloride
C36H30Cl2P2Pd
685607
Bis(triphenylphosphine)palladium(II) dichloride, ChemDose tablets, Loading: 1μmol per tablet
C36H30Cl2P2Pd
651745
Copper(I) chloride, anhydrous, beads, ≥99.99% trace metals basis
ClCu
229628
Copper(I) chloride, ≥99.995% trace metals basis
ClCu
224332
Copper(I) chloride, ReagentPlus®, purified, ≥99%
ClCu
212946
Copper(I) chloride, reagent grade, 97%
ClCu
61168
Copper(I) chloride, puriss. p.a., ACS reagent, ≥97.0% (RT)
ClCu
61169
Copper(I) chloride, purum, ≥97.0% (RT)
ClCu
03140
Copper(I) iodide, purum, ≥99.5%
CuI
215554
Copper(I) iodide, 99.999% trace metals basis
CuI
205540
Copper(I) iodide, 98%
CuI
792063
Copper(I) iodide, anhydrous, 99.995% trace metals basis
CuI
93092
Triphenylphosphine, ≥95.0% (GC)
C18H15P
T84409
Triphenylphosphine, ReagentPlus®, 99%
C18H15P
14664
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~3.2 mmol/g loading
C18H15P
93093
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~3 mmol/g triphenylphosphine loading
C18H15P
366455
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~3.0 mmol/g loading, 2 % cross-linked with divinylbenzene
C18H15P
93094
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~1.6 mmol/g loading
C18H15P