EMAIL THIS PAGE TO A FRIEND
Malaria Journal
Colonization of malaria vectors under semi-field conditions as a strategy for maintaining genetic and phenotypic similarity with wild populations.
PMID 25604997