EMAIL THIS PAGE TO A FRIEND
The Journal of infectious diseases
A thermonuclease of Neisseria gonorrhoeae enhances bacterial escape from killing by neutrophil extracellular traps.
PMID 25605868
Abstract
Acute gonorrhea is characterized by neutrophilic inflammation that is insufficient to clear Neisseria gonorrhoeae. Activated neutrophils release extracellular traps (NETs), which are composed of chromatin and decorated with antimicrobial proteins. The N. gonorrhoeae NG0969 open reading frame contains a gene (nuc) that encodes a putatively secreted thermonuclease (Nuc) that contributes to biofilm remodeling. Here, we report that Nuc degrades NETs to help N. gonorrhoeae resist killing by neutrophils. Primary human neutrophils released NETs after exposure to N. gonorrhoeae, but NET integrity declined over time with Nuc-containing bacteria. Recombinant Nuc and conditioned medium from Nuc-containing N. gonorrhoeae degraded human neutrophil DNA and NETs. NETs were found to have antimicrobial activity against N. gonorrhoeae, and Nuc expression enhanced N. gonorrhoeae survival in the presence of neutrophils that released NETs. We propose that Nuc enables N. gonorrhoeae to escape trapping and killing by NETs during symptomatic infection, highlighting Nuc as a multifunctional virulence factor for N. gonorrhoeae.
Related Materials