EMAIL THIS PAGE TO A FRIEND
Biochemical Society transactions
Effect of glucose analogues on yeast adenylate cyclase in vitro.
PMID 2560728
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9761
6-Deoxy-D-glucose
C6H12O5