EMAIL THIS PAGE TO A FRIEND
Dna and Cell Biology
The traditional Chinese medicinal formula BDL301 suppresses tumor growth by inhibiting STAT3 pathway and inducing apoptosis in colorectal cancer cells.
PMID 25607688