EMAIL THIS PAGE TO A FRIEND
The Journal of Clinical Investigation
Disrupting SUMOylation enhances transcriptional function and ameliorates polyglutamine androgen receptor-mediated disease.
PMID 25607844