EMAIL THIS PAGE TO A FRIEND
International journal of molecular sciences
Unravelling genes and pathways implicated in working memory of schizophrenia in Han Chinese.
PMID 25608650
Abstract
Working memory deficit is the core neurocognitive disorder in schizophrenia patients. To identify the factors underlying working memory deficit in schizophrenia patients and to explore the implication of possible genes in the working memory using genome-wide association study (GWAS) of schizophrenia, computerized delay-matching-to-sample (DMS) and whole genome genotyping data were obtained from 100 first-episode, treatment-naïve patients with schizophrenia and 140 healthy controls from the Mental Health Centre of the West China Hospital, Sichuan University. A composite score, delay-matching-to-sample total correct numbers (DMS-TC), was found to be significantly different between the patients and control. On associating quantitative DMS-TC with interactive variables of groups × genotype, one SNP (rs1411832), located downstream of YWHAZP5 in chromosome 10, was found to be associated with the working memory deficit in schizophrenia patients with lowest p-value (p = 2.02 × 10(-7)). ConsensusPathDB identified that genes with SNPs for which p values below the threshold of 5 × 10(-5) were significantly enriched in GO:0007155 (cell adhesion, p < 0.001). This study indicates that working memory, as an endophenotype of schizophrenia, could improve the efficacy of GWAS in schizophrenia. However, further study is required to replicate the results from our study.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
76048
Dimethyl sulfide, analytical standard
C2H6S