EMAIL THIS PAGE TO A FRIEND
Nature communications
A proteomic approach reveals integrin activation state-dependent control of microtubule cortical targeting.
PMID 25609142
Abstract
Integrin activation, which is regulated by allosteric changes in receptor conformation, enables cellular responses to the chemical, mechanical and topological features of the extracellular microenvironment. A global view of how activation state converts the molecular composition of the region proximal to integrins into functional readouts is, however, lacking. Here, using conformation-specific monoclonal antibodies, we report the isolation of integrin activation state-dependent complexes and their characterization by mass spectrometry. Quantitative comparisons, integrating network, clustering, pathway and image analyses, define multiple functional protein modules enriched in a conformation-specific manner. Notably, active integrin complexes are specifically enriched for proteins associated with microtubule-based functions. Visualization of microtubules on micropatterned surfaces and live cell imaging demonstrate that active integrins establish an environment that stabilizes microtubules at the cell periphery. These data provide a resource for the interrogation of the global molecular connections that link integrin activation to adhesion signalling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
755842
6-Maleimidohexanoic acid, 90% (GC)
C10H13NO4
63176
6-Maleimidohexanoic acid, ≥98.0% (HPLC)
C10H13NO4