EMAIL THIS PAGE TO A FRIEND
Luminescence : the journal of biological and chemical luminescence
Micelle-enhanced spectrofluorimetric determination of amlexanox in bioadhesive buccal tablets: application to content uniformity testing.
PMID 25611457
Abstract
A highly sensitive, simple and rapid spectrofluorimetric method was developed for the determination of Amlexanox (AMX) in its bioadhesive buccal tablets. The proposed method is based on measuring the native fluorescence of the methanolic solution of AMX at 400 nm after excitation at 242 nm in 0.2 M borate buffer (pH 10) and 0.5% w/v sodium dodecyl sulfate (SDS) solution. The interaction of AMX with SDS was studied, and the enhanced fluorescence intensity was exploited to develop an assay method for the determination of AMX. The relative fluorescence intensity-concentration plot was rectilinear over the range 5.0-80.0 ng/mL, with a lower detection limit of 0.57 ng/mL and a lower quantification limit of 1.74 ng/mL. The proposed method was successfully applied to the analysis of AMX in its commercial tablets. Moreover, content uniformity testing was conducted by applying official USP guidelines. Statistical evaluation and comparison of the data obtained using the proposed and comparison methods revealed good accuracy and precision for the proposed method.