EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Development and validation of determinative and confirmatory LC-MS/MS methodologies for total florfenicol and tulathromycin residues in bovine, equine and porcine kidney, liver and muscle tissues.
PMID 25612770
Abstract
Separate methods for the quantitation and confirmation of regulatory relevant residue concentrations of total florfenicol and tulathromycin residues in multiple tissue matrices were developed and validated. Total florfenicol residues, determined and expressed as florfenicol amine (FFA) equivalents, were quantified and confirmed over a concentration range of 100-4000ng/g, with an LOD of 33ng/g, while total tulathromyicn residues, determined as CP-60,300 and expressed as tulathromycin equivalents, were quantified and confirmed over a concentration range of 500-10,000ng/g, with an LOD of 300ng/g. A 2 or 1h acid digestion for the FFA and tulathromycin methods, respectively, followed by extraction, cleanup, and concentration using mixed-mode strong cation-exchange SPE cartridges was used. Quantitation and confirmation were accomplished using commercially available tri-deuterated FFA (FFA-D3) as internal standard and multiple reaction monitoring (MRM) of three transitions per target analyte. Mean recoveries and matrix effects were 60% and 25%; and 100% and 110%, respectively for the target analytes florfenicol amine and CP-60,300. Intra-lab method reproducibilities ranged from 7 to 11% RSD, which are within the AOAC recommended HORRATr guidelines for method reproducibilities estimated from single laboratory validation studies. Blind spikes showed that method bias was generally less than 15% for both methods within the calibration range. Both methods have been shown to meet requirements for use in national chemical residue monitoring programs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32492
Florfenicol amine, VETRANAL, analytical standard
C10H14FNO3S