EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Isolation of aspalathin and nothofagin from rooibos (Aspalathus linearis) using high-performance countercurrent chromatography: sample loading and compound stability considerations.
PMID 25614190