EMAIL THIS PAGE TO A FRIEND
Biologicals : Journal of the International Association of Biological Standardization
The importance of formulation in the successful lyophilization of influenza reference materials.
PMID 25614372