EMAIL THIS PAGE TO A FRIEND
Current opinion in cell biology
Extracellular signals and intracellular transduction pathways regulating Dictyostelium development.
PMID 2561454
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0567
DIF-3, ≥99% (HPLC), solid
C13H17ClO4