EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Detoxification of the veterinary antibiotic chloramphenicol using electron beam irradiation.
PMID 25616384
Abstract
Electron beam irradiation has shown potential as an alternative process for the treatment of industrial effluents that contain toxic organic chemicals. This study investigated the effectiveness of electron beam in degrading chloramphenicol (CAP) in aqueous solution. The degradation efficiency was 32.4% at 1 kGy, 86.9% at 5 kGy, and 100% at 10 kGy. The total organic carbon (TOC) of CAP in aqueous solution declined 4.6% at 1 kGy, 12.1% at 5 kGy, and 17.1% at 10 kGy of irradiation with electron beam. The CAP degradation products after irradiation were CAP1 ([M + H] m/z 307.1), CAP2 ([M + H] m/z 291.1), and CAP3 ([M + H] m/z 321.1). The degradation products were tested for microbial toxicity against Escherichia coli, Pseudomonas putida, and Bacillus subtilis and did not show any toxic antimicrobial effects caused by the CAP degradation products after irradiation with electron beam. The results of this study suggest that electron beam irradiation is the best technology for the comprehensive treatment of veterinary antibiotics at wastewater treatment plants.