EMAIL THIS PAGE TO A FRIEND
Journal of Chromatographic Science
Simultaneous Determination of Tramadol and Its Metabolite in Human Urine by the Gas Chromatography-Mass Spectrometry Method.
PMID 25616987
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
UC304
O-Desmethyltramadol
C15H23NO2
42965
Tramadol hydrochloride, ≥99.0% (HPLC)
C16H25NO2 · HCl
UC456
Tramadol hydrochloride, ≥98% (HPLC), solid
C16H25NO2 · HCl