EMAIL THIS PAGE TO A FRIEND
Biomaterials
Prolonged prevention of retinal degeneration with retinylamine loaded nanoparticles.
PMID 25617130
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S4000
6-Selenopurine
C5H4N4Se