EMAIL THIS PAGE TO A FRIEND
Journal of Pharmaceutical and Biomedical Analysis
Achiral-chiral two-dimensional chromatography of free amino acids in milk: A promising tool for detecting different levels of mastitis in cows.
PMID 25617178