EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology. Lung Cellular and Molecular Physiology
Prevention of pulmonary hypoplasia and pulmonary vascular remodeling by antenatal simvastatin treatment in nitrofen-induced congenital diaphragmatic hernia.
PMID 25617377
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33374
Nitrofen, PESTANAL®, analytical standard
C12H7Cl2NO3