EMAIL THIS PAGE TO A FRIEND
The Journal of Experimental Biology
Reproduction potentiated in nematodes (Caenorhabditis elegans) and guppy fish (Poecilia reticulata) by adding a synthetic peptide to their aqueous environment.
PMID 25617450