EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Development of a more specific and accurate multiple reaction monitoring method based on GC-EI/MS/MS for simultaneously monitoring and determining 34 kinds of pesticides in Qianjinzhidai pills.
PMID 25618250
Abstract
A rapid and accurate multi-residue method was developed for simultaneously monitoring and determining 34 kinds of pesticides by GC-EI/MS/MS, successfully applied in Qianjinzhidai pills. Precision, repeatability, stability, limit of detection (LOD), limit of quantification (LOQ), linearity range and accuracy tests were used to validate this method. The results, precision (RSD<2%, n=6), repeatability (RSD<2%, n=6) and stability (RSD<2%, n=6), LOD (lower than 0.02mg/kg), LOQ (lower than 0.067mg/kg), linearity range (0.016-39.270μg/mL), recoveries (90-110%, RSD<15%, n=6), indicated that the method was feasible. This paper provided an optimization method about ion transitions which can improve the specificity of mass spectrum monitoring and the accuracy of content determination. This method can be also used to monitor and determine pesticide residues of other traditional Chinese medicine, but it was worth noting that the ion transitions should been again optimized according to the sample matrix.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45315
Acephate, PESTANAL®, analytical standard
C4H10NO3PS
SCC-003
cis-Chlordane, vial of 1 g, (α isomer), analytical standard
C10H6Cl8
45428
Diazinon, PESTANAL®, analytical standard
C12H21N2O3PS
68486
Diazinon, certified reference material, TraceCERT®
C12H21N2O3PS
33491
Dieldrin, PESTANAL®, analytical standard
C12H8Cl6O
SCD-004
Dieldrin, vial of 1 g, 99%, analytical standard
C12H8Cl6O
36660
Dieldrin solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
C12H8Cl6O
40088
Dieldrin solution, certified reference material, 1000 μg/mL in methanol
C12H8Cl6O
45449
Dimethoate, PESTANAL®, analytical standard
C5H12NO3PS2
36676
Endosulfan sulfate, PESTANAL®, analytical standard
C9H6Cl6O4S
45477
Ethion, PESTANAL®, analytical standard
C9H22O4P2S4
32349
Heptachlor, PESTANAL®, analytical standard
C10H5Cl7
N12147
Heptachlor, analytical standard
C10H5Cl7
45522
Hexachlorobenzene, PESTANAL®, analytical standard
C6Cl6
48508
Hexachlorobenzene, analytical standard
C6Cl6
N12393
Methamidophos, analytical standard
C2H8NO2PS
33395
Methamidophos, PESTANAL®, analytical standard
C2H8NO2PS
PS676
Methamidophos, analytical standard
C2H8NO2PS
N12397
Methidathion, analytical standard
C6H11N2O4PS3
36158
Methidathion, PESTANAL®, analytical standard
C6H11N2O4PS3
PS679
Methidathion, analytical standard
C6H11N2O4PS3
36181
Omethoate, PESTANAL®, analytical standard
C5H12NO4PS
45607
Parathion, PESTANAL®, analytical standard
C10H14NO5PS
49062
Parathion, analytical standard
C10H14NO5PS
P8556
Pentachloronitrobenzene, plant cell culture tested, BioReagent, ≥94% (GC), powder
C6Cl5NO2
P2205
Pentachloronitrobenzene, ≥94%
C6Cl5NO2
45653
Quintozene, PESTANAL®, analytical standard
C6Cl5NO2
SCC-004
trans-Chlordane (gamma-chlordane), vial of 1 g, (γ isomer), analytical standard
C10H6Cl8
ERC-004
trans-Chlordane (gamma-chlordane), (γ isomer), vial of 25 mg, analytical standard
C10H6Cl8