EMAIL THIS PAGE TO A FRIEND
Archives of gynecology and obstetrics
Prophylactic vitamin D supplementation in ovarian hyperstimulation syndrome: an animal study.
PMID 25618750
Abstract
To investigate the effect of vitamin D in ovarian hyperstimulation syndrome (OHSS). In this animal study, 28 immature female Wistar rats were divided into four groups: group 1 (control); group 2 (ovarian stimulation); group 3 (OHSS group); group 4 (OHSS + vitamin D group). All groups were killed 48 h after hCG administration and were compared in terms of vascular permeability, ovarian weight, ovarian diameter, vascular endothelial growth factor (VEGF) expression (immunohistochemistry) in ovarian tissue and pigment epithelium-derived factor (PEDF) level in the serum (ELISA test) with the Kruskal-Wallis and Mann-Whitney U tests. VEGF expression in the vitamin D group was similar to that in the OHSS group. However, the PEDF level was significantly higher in the vitamin D group (p = 0.013). Prophylactic vitamin D supplementation is not sufficiently effective in preventing OHSS. Vitamin D effectively increases PEDF, which has an opposing effect on VEGF, which plays a key role in OHSS. Thus, the protective effect of Vitamin D on OHSS should be investigated with a vitamin D deficient model in the study group.