EMAIL THIS PAGE TO A FRIEND
Analytical and Bioanalytical Chemistry
Application of hydrostatic CCC-TLC-HPLC-ESI-TOF-MS for the bioguided fractionation of anticholinesterase alkaloids from Argemone mexicana L. roots.
PMID 25618762