EMAIL THIS PAGE TO A FRIEND
Acta Pharmacologica Sinica
Matrine pretreatment improves cardiac function in rats with diabetic cardiomyopathy via suppressing ROS/TLR-4 signaling pathway.
PMID 25619390
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11038
2,3-Butanedione, analytical standard
C4H6O2
B85307
2,3-Butanedione, 97%
C4H6O2