EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences
A validated UPLC-MS/MS assay using negative ionization mode for high-throughput determination of pomalidomide in rat plasma.
PMID 25621435
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0018
Pomalidomide, ≥98% (HPLC)
C13H11N3O4