EMAIL THIS PAGE TO A FRIEND
The International Journal of Biochemistry & Cell Biology
The PDZ3 domain of the cellular scaffolding protein MAGI-1 interacts with the Coxsackievirus and adenovirus receptor (CAR).
PMID 25622559