EMAIL THIS PAGE TO A FRIEND
Journal of Pineal Research
Melatonin sensitizes human breast cancer cells to ionizing radiation by downregulating proteins involved in double-strand DNA break repair.
PMID 25623566