EMAIL THIS PAGE TO A FRIEND
The Journal of Experimental Medicine
PI3-kinase activation is critical for host barrier permissiveness to Listeria monocytogenes.
PMID 25624443
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35441
Wortmannin solution, 100 μg/mL in acetonitrile, analytical standard
C23H24O8