EMAIL THIS PAGE TO A FRIEND
Drug design, development and therapy
Alisertib, an Aurora kinase A inhibitor, induces apoptosis and autophagy but inhibits epithelial to mesenchymal transition in human epithelial ovarian cancer cells.
PMID 25624750