EMAIL THIS PAGE TO A FRIEND
Oncology letters
CD26-positive/CD326-negative circulating cancer cells as prognostic markers for colorectal cancer recurrence.
PMID 25624884
Abstract
The present study evaluated the presence and clinical relevance of a cluster of differentiation (CD)26