EMAIL THIS PAGE TO A FRIEND
The New phytologist
Silicon induces resistance to the brown spot fungus Cochliobolus miyabeanus by preventing the pathogen from hijacking the rice ethylene pathway.
PMID 25625327
Abstract
Although numerous studies have shown the ability of silicon (Si) to mitigate a wide variety of abiotic and biotic stresses, relatively little is known about the underlying mechanism(s). Here, we have investigated the role of hormone defense pathways in Si-induced resistance to the rice brown spot fungus Cochliobolus miyabeanus. To delineate the involvement of multiple hormone pathways, a multidisciplinary approach was pursued, combining exogenous hormone applications, pharmacological inhibitor experiments, time-resolved hormone measurements, and bioassays with hormone-deficient and/or -insensitive mutant lines. Contrary to other types of induced resistance, we found Si-induced brown spot resistance to function independently of the classic immune hormones salicylic acid and jasmonic acid. Our data also rule out a major role of the abscisic acid (ABA) and cytokinin pathways, but suggest that Si mounts resistance to C.xa0miyabeanus by preventing the fungus from hijacking the rice ethylene (ET) machinery. Interestingly, rather than suppressing rice ET signaling per se, Si probably interferes with the production and/or action of fungal ET. Together our findings favor a scenario whereby Si induces brown spot resistance by disarming fungal ET and argue that impairment of pathogen virulence factors is a core resistance mechanism underpinning Si-induced plant immunity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3903
1-Aminocyclopropanecarboxylic acid, ≥98% (TLC)
C4H7NO2
36759
2,2′-Bipyridyl, PESTANAL®, analytical standard
C10H8N2
D216305
2,2′-Bipyridyl, ReagentPlus®, ≥99%
C10H8N2
B3408
6-Benzylaminopurine, plant cell culture tested
C12H11N5
13151
6-Benzylaminopurine, ReagentPlus®, ≥99.0% (HPLC)
C12H11N5
67619
6-Benzylaminopurine, analytical standard
C12H11N5
409332
Cobalt(II) chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Cl2Co
449776
Cobalt(II) chloride, anhydrous, beads, −10 mesh, 99.9% trace metals basis
Cl2Co
C0143
Ethephon, ≥96% (titration)
C2H6ClO3P
45473
Ethephon, PESTANAL®, analytical standard
C2H6ClO3P
G7645
Gibberellic acid, plant cell culture tested, BioReagent, ≥90% gibberellin A3 basis (of total gibberellins.)
C19H22O6
48880
Gibberellic acid, 90% gibberellin A3 basis (HPLC)
C19H22O6
36575
Gibberellic acid, PESTANAL®, analytical standard
C19H22O6
G1025
Gibberellic acid potassium salt, BioReagent, plant cell culture tested, ~95%, ≥50% total GA3 basis
C19H21KO6
48870
Gibberellin, 80% gibberellin A3 basis (TLC)
C19H22O6
CDS014278
N-benzyl-9H-purin-6-amine, AldrichCPR
C12H11N5
SIL350
Silicic acid, BioReagent, suitable for column chromatography, 100-200 mesh (75 - 150 μm)
H2O3Si
SILAW
Silicic acid, BioReagent, suitable for column chromatography, 200-400 mesh (38 - 75 μm)
H2O3Si
SILA200
Silicic acid, suitable for column chromatography, 60-200 mesh
H2O3Si
S7026
Sodium thiosulfate
Na2O3S2
563188
Sodium thiosulfate, ≥99.99% trace metals basis
Na2O3S2
HT1005
Sodium thiosulfate solution, 2 g/dL in deionized water
Na2O3S2
Z0876
trans-Zeatin, BioReagent, plant cell culture tested, ≥97%
C10H13N5O
Z2753
trans-Zeatin hydrochloride, plant cell culture tested, ≥97%
C10H13N5O · HCl