EMAIL THIS PAGE TO A FRIEND
Sensors (Basel, Switzerland)
Regeneration of recombinant antigen microarrays for the automated monitoring of antibodies against zoonotic pathogens in swine sera.
PMID 25625908
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P3786
Potassium phosphate dibasic, ACS reagent, ≥98%
HK2O4P