EMAIL THIS PAGE TO A FRIEND
Journal of Proteome Research
Phospholipid profile of amniotic fluid in ovine model of congenital diaphragmatic hernia (CDH): the effect of fetal tracheal occlusion.
PMID 25625961