EMAIL THIS PAGE TO A FRIEND
Revue medicale suisse
[Are we uro-ecologists?].
PMID 25626253
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295396
Hydrogen, ≥99.99%
H2