EMAIL THIS PAGE TO A FRIEND
Nihon rinsho. Japanese journal of clinical medicine
[Treatment of multiple myeloma with lenalidomide and bortezomib combination therapy].
PMID 25626318
Abstract
In the past decade, the introduction of bortezomib, thalidomide, and lenalidomide has changed the treatment of multiple myeloma (MM) dramatically. Combinations of bortezomib or lenalidomide with dexamethasone have improved response rates and prolonged response duration compared with older approaches. However, many patients will relapse, and efforts to improve survival are still needed. Several clinical studies have demonstrated that bortezomib- or lenalidomide-based triplet regimens have improved activity compared with doublet regimens. Bortezomib and lenalidomide have different but overlapping mechanisms of anti-MM activity in preclinical studies. In several studies, the triplet regimens of lenalidomide, bortezomib, and dexamethasone (RVD) have shown significant efficacy and favorable tolerability in both newly diagnosed and relapsed/refractory multiple myeloma. In this review, we will focus on RVD therapy for myeloma treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T144
(±)-Thalidomide, ≥98%, powder
C13H10N2O4