EMAIL THIS PAGE TO A FRIEND
Revista da Sociedade Brasileira de Medicina Tropical
Hepatitis B virus genotypes and mutations in the basal core promoter and pre-core/core in chronically infected patients in southern Brazil: a cross-sectional study of HBV genotypes and mutations in chronic carriers.
PMID 25626648
Abstract
In Brazil, little data exist regarding the distribution of genotypes in relation to basal core promoter (BCP) and precore/core mutations among chronic hepatitis B virus (HBV) carriers from different regions of the country. The aim of this study was to identify HBV genotypes and the frequency of mutations at the BCP and precore/core region among the prevalent genotypes in chronic carriers from southern Brazil. Nested-polymerase chain reaction (nested-PCR) products amplified from the S-polymerase gene, BCP and precore/core region from 54 samples were sequenced and analyzed. Phylogenetic analysis of the S-polymerase gene sequences showed that 66.7% (36/54) of the patients were infected with genotype D (D1, D2, D3), 25.9% (14/54) with genotype A (A1, A2), 5.6% (3/54) with subgenotype C2, and 2% (1/54) with genotype E. A comparison of virological characteristics showed significant differences between genotypes A, C and D. The comparison between HBeAg status and the G1896A stop codon mutation in patients with genotype D revealed a relationship between HBV G1896A precore mutants and genotype D and hepatitis B e antigen (HBeAg) seroconversion. Genotype D had a higher prevalence of the G1896A mutation and the presence of a thymine at position 1858. Genotype A was associated with a higher prevalence of the G1862T mutation and the presence of a cytosine at position 1858. HBV genotype D (D3) is predominant in HBV chronic carriers from southern Brazil. The presence of mutations in the BCP and precore/core region was correlated with the HBV genotype and HBeAg negative status.