EMAIL THIS PAGE TO A FRIEND
Bioprocess and biosystems engineering
Influence of oxygen on lovastatin biosynthesis by Aspergillus terreus ATCC 20542 quantitatively studied on the level of individual pellets.
PMID 25627471
Abstract
Despite oxygen is believed to be the most important environmental factor for any aerobic microbial process, the quantitative studies of its influence on growth and metabolite formation on the level of individual pellets formed by filamentous fungi were seldom performed. Never was it made for lovastatin producer Aspergillus terreus ATCC20542. Thus, this work is a quantitative study of oxygen transfer into A. terreus pellets during lovastatin biosynthesis in the shake flask culture. The basic measurement tool was an oxygen microprobe allowing for obtaining oxygen concentration profiles in the pellets. The pellets of various sizes from 1,600 to 6,400 μm exerting different oxygen transfer conditions were studied. Also various initial concentrations of carbon source were applied to change the conditions of biological reaction running in the pellets. Effective diffusivities in A. terreus pellets ranged from 643 to 1,342 μm s(-1) dependent on their size and structure. It occurred that only the smallest pellets of diameter equal to about 1,400 μm were fully penetrated by oxygen. What is more, apart from the size of pellets, the appropriate lactose concentration was required to effectively produce lovastatin. Its value was correlated with oxygen concentration on the surface of the pellet and could not be either too high, as the aforementioned oxygen level tended then to zero, or too low, as despite high oxygen concentration no biological reaction ran in the pellet and no lovastatin was formed.