EMAIL THIS PAGE TO A FRIEND
Scientific Reports
Apoptosis signal-regulating kinase 1 promotes Ochratoxin A-induced renal cytotoxicity.
PMID 25627963
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295604
Oxygen, ≥99.6%
O2