EMAIL THIS PAGE TO A FRIEND
Journal of translational medicine
NADPH oxidase p47phox siRNA attenuates adventitial fibroblasts proliferation and migration in apoE(-/-) mouse.
PMID 25628043
Abstract
Reactive oxide species (ROS) derived from NADPH oxidases is involved in atherosclerosis. However, as a key component of NADPH oxidase, how p47phox regulates NADPH oxidases activity, ROS production and adventitial fibroblasts (AFs) function remains unclear. p47phox in aortic arteries of apoE(-/-) mice fed with hyperlipid diet was detected by immunohistochemistry. NADPH oxidase activity, superoxide anion (O2(-)) generation and p47phox expression were analyzed in primary AFs treated by diphenyleneiodonium (DPI). The proliferation and migration of AFs were also analyzed. p47phox expression was low in the aortic adventitia but high in the site of intimal injury with continuous hyperlipidic diet. Compared to AFs from wild-type mice, AFs derived from apoE(-/-) mice exhibited elevated NADPH oxidase activity, O2(-) production and higher mRNA and protein levels of p47phox, correlated with increased capability of proliferation and migration. DPI inhibited NADPH oxidase activity and AFs proliferation and migration in a dose-dependent manner. In addition, siRNA mediated knockdown of p47phox attenuated the proliferation and migration of AFs derived from apoE(-/-) mice. p47phox plays a critical role in the regulation of adventitial fibroblast proliferation and migration and may be a new therapeutic target for neointimal hyperplasia.